missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Product Code. 10707533 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10707533

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35138-22-8 | C16H24BF4Rh | 406.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35138-22-8
Molecular Formula C16H24BF4Rh
Molecular Weight (g/mol) 406.08
MDL Number MFCD00075045
InChI Key NBGSCPNNKGVTKU-QMDOQEJBSA-N
Synonym bis 1,5-cyclooctadiene rhodium i tetrafluoroborate, rhodium i tetrafluoroborate 1,5-cyclooctadiene complex, 1z,5z-cycloocta-1,5-diene; rhodium; tetrafluoroborate, bis 1,5-cyclooctadiene rhodium tetrafluoroborate, rh cod 2 bf4, c16h24bf4rh, c16h24rh.bf4, bis 1,5-cyclooctadien rhodium i tetrafluoroborate
PubChem CID 5702662
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;rhodium;tetrafluoroborate
SMILES [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 210.0°C
Color Red
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula [Rh(cod)2]BF4
Quantity 250 mg
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons
Formula Weight 406.08
Physical Form Powder
Chemical Name or Material Bis(1, 5-Cyclooctadiene)rhodium(I) tetrafluoroborate
compliance-icons compliance-icons
Product Identifier
  • Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.