missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11364068
Click to view available options
Quantity:
0.5 g
2 g
10 g

Precursor for CVD process

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II) is used in nano-material, thin films and ferroelectric metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13986-53-3
Molecular Formula C22H38CoO4
Molecular Weight (g/mol) 425.48
MDL Number MFCD00233616
InChI Key KLJJOSZRALJWDS-UHFFFAOYSA-N
Synonym bis 2,2,6,6-tetramethyl-3,5-heptanedionato cobalt ii, cobalt 2+ bis 2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
PubChem CID 57369563
IUPAC Name cobalt(2+);2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
SMILES [Co++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

Specifications

Melting Point 143°C
Quantity 10 g
Solubility Information Soluble in hydrocarbons,alcohols and ethers. Slightly soluble in water.
Formula Weight 425.46
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'