Learn More
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis), Thermo Scientific Chemicals
Precursor for CVD process
£42.90 - £546.00
Chemical Identifiers
CAS | 13986-53-3 |
---|---|
Molecular Formula | C22H38CoO4 |
Molecular Weight (g/mol) | 425.48 |
MDL Number | MFCD00233616 |
InChI Key | KLJJOSZRALJWDS-UHFFFAOYSA-N |
Synonym | bis 2,2,6,6-tetramethyl-3,5-heptanedionato cobalt ii, cobalt 2+ bis 2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate |
PubChem CID | 57369563 |
IUPAC Name | cobalt(2+);2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate |
SMILES | [Co++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384068
|
Thermo Scientific Alfa Aesar
041359.02 |
0.5 g |
£42.90
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11374068
|
Thermo Scientific Alfa Aesar
041359.04 |
2 g |
£146.00
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11364068
|
Thermo Scientific Alfa Aesar
041359.09 |
10 g |
£546.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II) is used in nano-material, thin films and ferroelectric metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13986-53-3 | |
425.48 | |
KLJJOSZRALJWDS-UHFFFAOYSA-N | |
57369563 | |
[Co++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C |
C22H38CoO4 | |
MFCD00233616 | |
bis 2,2,6,6-tetramethyl-3,5-heptanedionato cobalt ii, cobalt 2+ bis 2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate | |
cobalt(2+);2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate |
Specifications
13986-53-3 | |
C22H38CoO4 | |
bis 2,2,6,6-tetramethyl-3,5-heptanedionato cobalt ii, cobalt 2+ bis 2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate | |
KLJJOSZRALJWDS-UHFFFAOYSA-N | |
cobalt(2+);2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate | |
57369563 | |
99.9% | |
(metals basis) |
143°C | |
MFCD00233616 | |
Soluble in hydrocarbons,alcohols and ethers. Slightly soluble in water. | |
[Co++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C | |
425.48 | |
425.46 | |
Powder | |
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II) |
Safety and Handling
GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H341
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only