missing translation for 'onlineSavingsMsg'
Learn More
Bis(2-diphenylphosphinophenyl)ether, 99%, Thermo Scientific Chemicals Product Code.: 10452483

Bis(2-diphenylphosphinophenyl)ether, 99%, Thermo Scientific Chemicals

Product Code. 10452483
25 g, Glass bottle
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10452483

Brand: Thermo Scientific Acros 383370250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166330-10-5
Molecular Formula C36H28OP2
Molecular Weight (g/mol) 538.57
MDL Number MFCD00233863
InChI Key RYXZOQOZERSHHQ-UHFFFAOYSA-N
Synonym bis 2-diphenylphosphinophenyl ether, dpephos, oxydi-2,1-phenylene bis diphenylphosphine, oxybis 2,1-phenylene bis diphenylphosphine, bis 2-diphenylphosphino phenyl ether, 2,2'-bis diphenylphosphino diphenyl ether, 2-2-diphenylphosphanyl phenoxy phenyl diphenylphosphane
PubChem CID 4285986
IUPAC Name [2-(2-diphenylphosphanylphenoxy)phenyl]-diphenylphosphane
SMILES O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 184.0°C to 187.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula [(C6H5)2P(C6H4)]2O
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 538.56
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Bis(2-diphenylphosphinophenyl)ether
Show More Show Less
compliance-icons
Product Identifier
  • Bis(2-diphenylphosphinophenyl)ether
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(2-diphenylphosphinophenyl)ether, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.