missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexyl)amine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Description
Bis(2-ethylhexyl)amine (BEA) was used in the synthesis of stable nanosized CdS particles coated by an oriented monolayer of chemically bonded BEA molecules
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bis(2-ethylhexyl)amine (BEA) was used in the synthesis of stable nanosized CdS particles coated by an oriented monolayer of chemically bonded BEA molecules
Solubility
Soluble in water 0.045 g/L @ 20˚C.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Bis(2-ethylhexyl)amine (BEA) was used in the synthesis of stable nanosized CdS particles coated by an oriented monolayer of chemically bonded BEA molecules
Solubility
Soluble in water 0.045 g/L @ 20˚C.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | <-60°C |
Density | 0.805 |
Boiling Point | 281°C |
Flash Point | 121°C (250°F) |
Refractive Index | 1.442 |
Quantity | 500 mL |
UN Number | UN2922 |
Beilstein | 1748342 |
Solubility Information | Soluble in water 0.045g/L @ 20?C. |
Formula Weight | 241.46 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Signal Word
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'