missing translation for 'onlineSavingsMsg'
Learn More
Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals Product Code.: 11925151

Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals

Product Code. 11925151
5 kg, Plastic drum
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 11925151

Brand: Thermo Scientific Acros 402470050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-07-7
Molecular Formula C16H35O4P
Molecular Weight (g/mol) 322.43
MDL Number MFCD00009492
InChI Key SEGLCEQVOFDUPX-UHFFFAOYNA-N
Synonym bis 2-ethylhexyl hydrogen phosphate, bis 2-ethylhexyl phosphate, hdehp, di 2-ethylhexyl phosphate, dehpa extractant, di 2-ethylhexyl phosphoric acid, escaid 100, d 2ehpa, bis 2-ethylhexyl phosphoric acidShow MoreShow Less
PubChem CID 9275
IUPAC Name bis(2-ethylhexyl) hydrogen phosphate
SMILES CCCCC(CC)COP(O)(=O)OCC(CC)CCCC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -60.0°C
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 48.0°C (12.0 mmHg)
Flash Point 137°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Plastic drum
Linear Formula (CH3(CH2)3CH(C2H5)CH2O)2POOH
Refractive Index 1.4400 to 1.4440
Quantity 5 kg
Beilstein 01,IV,1786
Specific Gravity 0.97
Solubility Information Solubility in water: slightly soluble
Formula Weight 322.42
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) hydrogen phosphate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459216 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog Number402470050,402470010
Lot NumberA0409872 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number402470050,402470010
Lot NumberA0409118 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number402470050,402470010
Lot NumberA0406724 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number402470050,402470010
Lot NumberA0418191 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number402470050,402470010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.