missing translation for 'onlineSavingsMsg'
Learn More
Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals Product Code.: 11373387

Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals

Product Code. 11373387
500 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11373387

Brand: Thermo Scientific Alfa Aesar 017723.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extraction of metals

Extraction of metals.Bis(2-ethylhexyl) phosphate is used as a lubricant additive, corrosion inhibitor and a metal extractant. It is involved in the solvent extraction of uranium salts and rare earth metals. Iron(II) ions are extracted after reduction of Iron(III) ions. Further, it is used as a plasticizer and as a solvent in the synthesis of plastic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-07-7
Molecular Formula C16H35O4P
Molecular Weight (g/mol) 322.43
MDL Number MFCD00009492
InChI Key SEGLCEQVOFDUPX-UHFFFAOYNA-N
Synonym bis 2-ethylhexyl hydrogen phosphate, bis 2-ethylhexyl phosphate, hdehp, di 2-ethylhexyl phosphate, dehpa extractant, di 2-ethylhexyl phosphoric acid, escaid 100, d 2ehpa, bis 2-ethylhexyl phosphoric acidShow MoreShow Less
PubChem CID 9275
IUPAC Name bis(2-ethylhexyl) hydrogen phosphate
SMILES CCCCC(CC)COP(O)(=O)OCC(CC)CCCC

Specifications

Melting Point -50°C
Density 0.965
Boiling Point 48°C (12 mmHg)
Flash Point 171°C (340°F)
Assay Percent Range 95%
Refractive Index 1.442
Quantity 500 g
UN Number 1902
Solubility Information Slightly miscible with water.
Formula Weight 322.43
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) Phosphate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU01K022 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog Number017723.22,017723.A3,017723.36
Lot NumberT27I021 Certificate TypeCertificate of Analysis Date04/08/2022 Catalog Number017723.22,017723.A3,017723.36
Lot NumberJ07M44 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number017723.22,017723.A3,017723.36
Lot NumberU04H005 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number017723.22,017723.A3,017723.36
Lot NumberC01S025 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number017723.22,017723.A3,017723.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.