missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-hydroxyethyl) disulfide, tech. 90%

Product Code. 11332627
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11332627

Brand: Thermo Scientific Alfa Aesar L11800.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1892-29-1 | C4H10O2S2 | 154.24 g/mol

Bis(2-hydroxyethyl) disulfide is used as an intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-hydroxyethyl) disulfide is used as an intermediate in chemical research.

Solubility
Soluble in water, ethanol, acetone, and ether.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1892-29-1
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00002906
InChI Key KYNFOMQIXZUKRK-UHFFFAOYSA-N
Synonym 2-hydroxyethyl disulfide, 2,2'-dithiodiethanol, bis 2-hydroxyethyl disulfide, ethanol, 2,2'-dithiobis, hydroxyethyl disulfide, diethanol disulfide, 2,2'-dithiobisethanol, 2-hydroxyethanedisulfide, usaf th-9, 2,2'-disulfanediyldiethanol
PubChem CID 15906
ChEBI CHEBI:43136
IUPAC Name 2-(2-hydroxyethyldisulfanyl)ethanol
SMILES OCCSSCCO

Specifications

Melting Point ∼25°C
Density 1.261
Boiling Point 158°C to 163°C (3.5 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Linear Formula HOCH2CH2S-SCH2CH2OH
Refractive Index 1.57
Quantity 250 g
UN Number UN2810
Beilstein 1735851
Solubility Information Soluble in water,ethanol,acetone,and ether.
Formula Weight 154.24
Percent Purity 90%
Grade Technical
Chemical Name or Material Bis(2-hydroxyethyl) disulfide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bis(2-hydroxyethyl) disulphide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Bis(2-hydroxyethyl)disulphide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.