Learn More
Bis(2-methoxyethyl) ether, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
£45.80 - £234.00
Chemical Identifiers
CAS | 111-96-6 |
---|---|
Molecular Formula | C6H14O3 |
Molecular Weight (g/mol) | 134.18 |
MDL Number | MFCD00008503 |
InChI Key | SBZXBUIDTXKZTM-UHFFFAOYSA-N |
Synonym | diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane |
PubChem CID | 8150 |
ChEBI | CHEBI:46784 |
IUPAC Name | 1-methoxy-2-(2-methoxyethoxy)ethane |
SMILES | COCCOCCOC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10500384
|
Thermo Scientific Acros
397211000 |
100 mL | AcroSeal™ Glass Bottle |
£45.80
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10590374
|
Thermo Scientific Acros
397210010 |
1 L | AcroSeal™ Glass Bottle |
£234.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-96-6 | |
134.18 | |
SBZXBUIDTXKZTM-UHFFFAOYSA-N | |
8150 | |
1-methoxy-2-(2-methoxyethoxy)ethane |
C6H14O3 | |
MFCD00008503 | |
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane | |
CHEBI:46784 | |
COCCOCCOC |
Specifications
111-96-6 | |
100.0 | |
Colorless | |
162.0°C | |
Authentic | |
C6H14O3 | |
1.4070 to 1.4090 | |
01, IV, 2393 | |
01,255 | |
0.937 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol,ether,hydrocarbon solvents | |
COCCOCCOC | |
134.18 | |
CHEBI:46784 | |
134.18 | |
Extra Dry | |
Bis(2-methoxyethyl) ether, Extra Dry |
99.0 | |
-64.0°C | |
0.9370g/mL | |
51°C | |
99% min. (GC) | |
(CH3OCH2CH2)2O | |
MFCD00008503 | |
0.001 max. | |
15, 3184 | |
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane | |
SBZXBUIDTXKZTM-UHFFFAOYSA-N | |
1-methoxy-2-(2-methoxyethoxy)ethane | |
8150 | |
2 mPa.s (20°C) | |
99+% | |
Liquid |
Safety and Handling
GHS H Statement
May damage fertility or the unborn child.
Flammable liquid and vapor.
May form explosive peroxides.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Use personal protective equipment as required.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-924-4
- Bis(2-methoxyethyl) ether
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
Hazard Statement
- H226-Flammable liquid and vapour.
- H360FD-May damage fertility. May damage the unborn child.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only