missing translation for 'onlineSavingsMsg'
Learn More

Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper(I) triflate, 97%, Thermo Scientific Chemicals

Product Code. 15465615
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874013-62-4
Molecular Formula C37H56CuF3O5P4S
Molecular Weight (g/mol) 857.346
MDL Number MFCD17018797
InChI Key JGVWSKIVCRMXGT-YCIFJGFLSA-M
Synonym bis 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-oxide benzene cu i trifluoromethanesulfonate
PubChem CID 11707733
IUPAC Name copper(1+);(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide;trifluoromethanesulfonate
SMILES CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.C(F)(F)(F)S(=O)(=O)[O-].[Cu+]

Specifications

Color Brown to White
Quantity 100 mg
Sensitivity Air Sensitive
Formula Weight 857.34
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'