missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate, Electronic grade, ≥99% (metals basis), Thermo Scientific Chemicals
£23.71 - £355.00
Chemical Identifiers
CAS | 131717-99-2 |
---|---|
Molecular Formula | C27H33IO3S |
Molecular Weight (g/mol) | 564.52 |
MDL Number | MFCD02683472 |
InChI Key | UEJFJTOGXLEPIV-UHFFFAOYSA-M |
Synonym | bis 4-tert-butylphenyl iodanium tosylate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, 4,4-di-tert-butyldiphenyliodonium p-toluenesulfonate, acmc-20n1fr, bis 4-tert-butylphenyl iodonium tosylate, bis 4-tert-butylphenl iodonium p, bis p-tert-butylphenyl iodonium p-toluenesulfonate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade trace metals basis |
PubChem CID | 16216932 |
IUPAC Name | bis(4-tert-butylphenyl)iodanium;4-methylbenzenesulfonate |
SMILES | CC1=CC=C(C=C1)S([O-])(=O)=O.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15485285
|
Thermo Scientific Alfa Aesar
H55188.MD |
250 mg | |||||||
15465285
|
Thermo Scientific Alfa Aesar
H55188.03 |
1 g |
£77.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15475285
|
Thermo Scientific Alfa Aesar
H55188.06 |
5 g |
£355.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
131717-99-2 | |
564.52 | |
UEJFJTOGXLEPIV-UHFFFAOYSA-M | |
16216932 | |
CC1=CC=C(C=C1)S([O-])(=O)=O.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1 |
C27H33IO3S | |
MFCD02683472 | |
bis 4-tert-butylphenyl iodanium tosylate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, 4,4-di-tert-butyldiphenyliodonium p-toluenesulfonate, acmc-20n1fr, bis 4-tert-butylphenyl iodonium tosylate, bis 4-tert-butylphenl iodonium p, bis p-tert-butylphenyl iodonium p-toluenesulfonate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade trace metals basis | |
bis(4-tert-butylphenyl)iodanium;4-methylbenzenesulfonate |
Specifications
131717-99-2 | |
White | |
MFCD02683472 | |
bis 4-tert-butylphenyl iodanium tosylate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, 4,4-di-tert-butyldiphenyliodonium p-toluenesulfonate, acmc-20n1fr, bis 4-tert-butylphenyl iodonium tosylate, bis 4-tert-butylphenl iodonium p, bis p-tert-butylphenyl iodonium p-toluenesulfonate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade trace metals basis | |
CC1=CC=C(C=C1)S([O-])(=O)=O.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1 | |
564.52 | |
564.52 | |
Electronic | |
(metals basis) |
237°C to 240°C | |
C27H33IO3S | |
Light sensitive | |
UEJFJTOGXLEPIV-UHFFFAOYSA-M | |
bis(4-tert-butylphenyl)iodanium;4-methylbenzenesulfonate | |
16216932 | |
≥99% | |
Powder | |
Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only