missing translation for 'onlineSavingsMsg'
Learn More

Bis(acetoxydimethyltin)oxide, Thermo Scientific™

Product Code. 11344547
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11344547

Brand: Thermo Scientific Alfa Aesar 039610.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5926-79-4
Molecular Formula C8H22O5Sn2
Molecular Weight (g/mol) 435.68
MDL Number MFCD00070453
InChI Key PTOQDQIJSRHODZ-UHFFFAOYSA-N
Synonym bis acetoxydimethyltin oxide, 1,3-diacetoxy-1,1,3,3-tetramethyldistannoxane, bis-acetoxydimethyltin oxide, acetyloxy dimethylstannyl oxy dimethylstannyl acetate
PubChem CID 16683964
IUPAC Name [[acetyloxy(dimethyl)stannyl]oxy-dimethylstannyl] acetate
SMILES O.C[SnH2]C.C[SnH2]C.CC(O)=O.CC(O)=O

Specifications

Melting Point 240°C
Quantity 25 g
UN Number UN3146
Sensitivity Moisture sensitive
Formula Weight 431.61
Physical Form Powder
Chemical Name or Material Bis(acetoxydimethyltin)oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.