missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(cyclopentadienyl)hafnium dichloride, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Starting material to prepare many kinds of hafnocene(II or IV) derivatives; in combination with AgI salt, is an effective activator for glycosyl fluoride. Pharmaceutical intermediates and alos used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Specifications
Melting Point | 230°C to 233°C |
Quantity | 1 g |
UN Number | UN3261 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in aromatic solvents,chloroform; slightly soluble in THF,ether. Insolble in hexanes. |
IUPAC Name | cyclopenta-1,3-diene;hafnium;dichloride |
Formula Weight | 379.59 |
Physical Form | Crystalline |
Chemical Name or Material | Bis(cyclopentadienyl)hafnium dichloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'