missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)hafnium dichloride, Thermo Scientific Chemicals

Product Code. 11343208
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Starting material to prepare many kinds of hafnocene(II or IV) derivatives; in combination with AgI salt, is an effective activator for glycosyl fluoride. Pharmaceutical intermediates and alos used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12116-66-4
Molecular Formula C10H10Cl2Hf
Molecular Weight (g/mol) 379.58
MDL Number MFCD00001438,MFCD00001438
InChI Key CSEGCHWAMVIXSA-UHFFFAOYSA-L
Synonym bis cyclopentadienyl hafniumdichloride
PubChem CID 131674741
SMILES [Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1

Specifications

Melting Point 230°C to 233°C
Quantity 25 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Solubility Information Soluble in aromatic solvents,chloroform; slightly soluble in THF,ether. Insolble in hexanes.
IUPAC Name hafnium(4+) bis(cyclopenta-2,4-dien-1-ide) dichloride
Formula Weight 379.59
Physical Form Crystalline
Chemical Name or Material Bis(cyclopentadienyl)hafnium dichloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'