Learn More
Bis(cyclopentadienyl)hafnium dichloride, Thermo Scientific Chemicals
£49.70 - £838.00
Chemical Identifiers
CAS | 12116-66-4 |
---|---|
Molecular Formula | C10H10Cl2Hf |
Molecular Weight (g/mol) | 379.58 |
MDL Number | MFCD00001438,MFCD00001438 |
InChI Key | CSEGCHWAMVIXSA-UHFFFAOYSA-L |
Synonym | bis cyclopentadienyl hafniumdichloride |
PubChem CID | 131674741 |
SMILES | [Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333208
|
Thermo Scientific Alfa Aesar
036105.03 |
1 g |
£49.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323208
|
Thermo Scientific Alfa Aesar
036105.06 |
5 g |
£177.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343208
|
Thermo Scientific Alfa Aesar
036105.14 |
25 g |
£838.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Starting material to prepare many kinds of hafnocene(II or IV) derivatives; in combination with AgI salt, is an effective activator for glycosyl fluoride. Pharmaceutical intermediates and alos used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
12116-66-4 | |
379.58 | |
CSEGCHWAMVIXSA-UHFFFAOYSA-L | |
131674741 |
C10H10Cl2Hf | |
MFCD00001438,MFCD00001438 | |
bis cyclopentadienyl hafniumdichloride | |
[Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1 |
Specifications
12116-66-4 | |
C10H10Cl2Hf | |
UN3261 | |
bis cyclopentadienyl hafniumdichloride | |
CSEGCHWAMVIXSA-UHFFFAOYSA-L | |
cyclopenta-1,3-diene;hafnium;dichloride | |
131674741 | |
Crystalline |
230°C to 233°C | |
MFCD00001438,MFCD00001438 | |
Air and moisture sensitive | |
Soluble in aromatic solvents,chloroform; slightly soluble in THF,ether. Insolble in hexanes. | |
[Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1 | |
379.58 | |
379.59 | |
Bis(cyclopentadienyl)hafnium dichloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 235-177-5
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only