missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)nickel, dry, Thermo Scientific Chemicals

Product Code. 15462977
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-28-9
Molecular Formula C10H10Ni
Molecular Weight (g/mol) 188.883
MDL Number MFCD00001441
InChI Key RQKPFSQDBAZFJV-UHFFFAOYSA-N
Synonym bis cyclopentadienyl nickel ii
PubChem CID 24942185
IUPAC Name cyclopenta-1,3-diene;nickel(2+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.[Ni+2]

Specifications

Melting Point 171°C to 173°C
Color Green
Quantity 50 g
UN Number UN3391
Sensitivity Air and moisture sensitive
Formula Weight 188.9
Physical Form Crystalline
Chemical Name or Material Bis (cyclopentadienyl)nickel, Dry

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'