Learn More
Bis(cyclopentadienyl)titanium dichloride, 97%, Thermo Scientific Chemicals
£128.00
Chemical Identifiers
CAS | 1271-19-8 |
---|---|
Molecular Formula | C10H10Cl2Ti |
Molecular Weight (g/mol) | 249 |
MDL Number | MFCD00003723 |
InChI Key | MKNXBRLZBFVUPV-UHFFFAOYSA-L |
Synonym | Titanocene dichloride |
PubChem CID | 124040768 |
IUPAC Name | cyclopenta-1,3-diene;titanium(2+);dichloride |
SMILES | [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1271-19-8 | |
249 | |
MKNXBRLZBFVUPV-UHFFFAOYSA-L | |
124040768 | |
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2] |
C10H10Cl2Ti | |
MFCD00003723 | |
Titanocene dichloride | |
cyclopenta-1,3-diene;titanium(2+);dichloride |
Specifications
1271-19-8 | |
100.0 | |
Bordeaux to Red | |
Authentic | |
C10H10Cl2Ti | |
06,48 | |
1.6 | |
Solubility in water: hydrolyses. Other solubilities: soluble in aromatic hydrocarbons, thf, halogenated, solvents, acetonitrile,, slightly soluble in chloroform, ethanol < 1mg/l,, acetone < 1mg/l | |
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2] | |
249 | |
249 | |
Crystalline Powder |
96.0 | |
287.0°C to 289.0°C | |
1.6000g/mL | |
96% min. (Argentometry) | |
MFCD00003723 | |
15, 9638 | |
Titanocene dichloride | |
MKNXBRLZBFVUPV-UHFFFAOYSA-L | |
cyclopenta-1,3-diene;titanium(2+);dichloride | |
124040768 | |
97% | |
Bis(cyclopentadienyl)titanium dichloride, 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Cal
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 215-035-9
missing translation for 'rtecsNumber' : XR2050000
missing translation for 'tsca' : TSCA
- Bis(cyclopentadienyl)titanium dichloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only