missing translation for 'onlineSavingsMsg'
Learn More

Bis(dibenzylideneacetone)palladium(0), Thermo Scientific Chemicals

Product Code. 11327547
Click to view available options
Quantity:
0.25 g
1 g
5 g

Hydrogenation, isomerization, carbonylation, oxidation, C-C bond formation.Bis(dibenzylideneacetone)palladium(0) is used as a reagent for the preparation of allylic substituted cyclopentadienes. It acts as a homogeneous catalyst as well as in the alkylation of allyl acetates by various nucleophiles. Further, it is used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32005-36-0
Molecular Formula C34H28O2Pd
Molecular Weight (g/mol) 575.02
MDL Number MFCD00051942
InChI Key UKSZBOKPHAQOMP-SVLSSHOZSA-N
Synonym bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
PubChem CID 6505921
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1

Specifications

Melting Point 135°C (decomposition)
Color Red to Brown
Quantity 5 g
Sensitivity Air and moisture sensitive
Solubility Information Slightly soluble in CH2Cl2,CHCl3 and benzene.Soluble in dichloromethane,chloroform,1,2-dichloroethane,acetone,acetonitrile and benzene. Insoluble in water.
Formula Weight 575
Physical Form Powder
Chemical Name or Material Bis(dibenzylideneacetone) palladium(0)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'