missing translation for 'onlineSavingsMsg'
Learn More

Bis(methylthio)methane, 99%, Thermo Scientific Chemicals

Product Code. 11448597
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1618-26-4
Molecular Formula C3H8S2
Molecular Weight (g/mol) 108.217
MDL Number MFCD00008564
InChI Key LOCDPORVFVOGCR-UHFFFAOYSA-N
Synonym bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3
PubChem CID 15380
IUPAC Name bis(methylsulfanyl)methane
SMILES CSCSC

Specifications

Boiling Point 146°C to 147°C
Flash Point 43°C (109°F)
Odor Unpleasant
Quantity 5 g
UN Number UN1993
Beilstein 1731143
Merck Index 14,1256
Formula Weight 108.23
Percent Purity 99%
Chemical Name or Material Bis(methylthio)methane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.