missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentafluorophenyl)phenylphosphine, 97%, Thermo Scientific Chemicals

Product Code. 11414493
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Bis(pentafluorophenyl)phenylphosphine is used as a ligand involved in the palladium-catalyzed Heck reaction of iodobenzene and styrene in compressed carbon dioxide. It is also used in the polymerization of 1,3-butadiene with Co(2-ethyl hexanoate)2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(pentafluorophenyl)phenylphosphine is used as a ligand involved in the palladium-catalyzed Heck reaction of iodobenzene and styrene in compressed carbon dioxide. It is also used in the polymerization of 1,3-butadiene with Co(2-ethyl hexanoate)2.

Solubility
Soluble in chloroform, dichloromethane, tetrahydrofuran, benzene, toluene, ethanol, methanol, and supercritical carbon dioxide.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5074-71-5
Molecular Formula C18H5F10P
Molecular Weight (g/mol) 442.20
MDL Number MFCD00000291
InChI Key OYNXPGGNQMSMTR-UHFFFAOYSA-N
Synonym bis pentafluorophenyl phenylphosphine, dftpp, decafluorotriphenylphosphine, bis perfluorophenyl phenyl phosphine, bis 2,3,4,5,6-pentafluorophenyl-phenylphosphane, phosphine,bis 2,3,4,5,6-pentafluorophenyl phenyl, bis 2,3,4,5,6-pentafluorophenyl phenyl phosphane, ultramark 443, bis pentafluorophenyl phenyl phosphite, bis pentafluorophenyl phenyl phosphine
PubChem CID 78757
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)-phenylphosphane
SMILES FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 59°C to 64°C
Boiling Point 105°C (0.3 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 6253897
Solubility Information Soluble in chloroform,dichloromethane,tetrahydrofuran,benzene,toluene,ethanol,methanol,and supercritical carbon dioxide.
Formula Weight 442.2
Percent Purity 97%
Chemical Name or Material Bis (pentafluorophenyl)phenylphosphine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'