missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(pentafluorophenyl)phenylphosphine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.25 g
1 g
Description
Bis(pentafluorophenyl)phenylphosphine is used as a ligand involved in the palladium-catalyzed Heck reaction of iodobenzene and styrene in compressed carbon dioxide. It is also used in the polymerization of 1,3-butadiene with Co(2-ethyl hexanoate)2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bis(pentafluorophenyl)phenylphosphine is used as a ligand involved in the palladium-catalyzed Heck reaction of iodobenzene and styrene in compressed carbon dioxide. It is also used in the polymerization of 1,3-butadiene with Co(2-ethyl hexanoate)2.
Solubility
Soluble in chloroform, dichloromethane, tetrahydrofuran, benzene, toluene, ethanol, methanol, and supercritical carbon dioxide.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids and bases.
Bis(pentafluorophenyl)phenylphosphine is used as a ligand involved in the palladium-catalyzed Heck reaction of iodobenzene and styrene in compressed carbon dioxide. It is also used in the polymerization of 1,3-butadiene with Co(2-ethyl hexanoate)2.
Solubility
Soluble in chloroform, dichloromethane, tetrahydrofuran, benzene, toluene, ethanol, methanol, and supercritical carbon dioxide.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids and bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 59°C to 64°C |
Boiling Point | 105°C (0.3 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 0.25 g |
Beilstein | 6253897 |
Solubility Information | Soluble in chloroform,dichloromethane,tetrahydrofuran,benzene,toluene,ethanol,methanol,and supercritical carbon dioxide. |
Formula Weight | 442.2 |
Percent Purity | 97% |
Chemical Name or Material | Bis (pentafluorophenyl)phenylphosphine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'