missing translation for 'onlineSavingsMsg'
Learn More
Bis(phenylacetyl) disulfide, 98%, Thermo Scientific Chemicals Product Code.: 11329755

Bis(phenylacetyl) disulfide, 98%, Thermo Scientific Chemicals

Product Code. 11329755
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11329755

Brand: Thermo Scientific Alfa Aesar L19792.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bis(phenylacetyl)disulfide as sulfur substitution agent in preparation of RNA oligonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(phenylacetyl)disulfide as sulfur substitution agent in preparation of RNA oligonucleotides.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15088-78-5
Molecular Formula C16H14O2S2
Molecular Weight (g/mol) 302.41
MDL Number MFCD00513572
InChI Key IXGZXXBJSZISOO-UHFFFAOYSA-N
Synonym phenylacetyl disulfide, 2-phenylacetic dithioperoxyanhydride, phenylacetyl disulphide, bis phenylacetyl disulfide, bis phenylacetyl disulphide, 2-phenyl-1-2-phenylacetyl disulfanyl ethanone, bis phenylacetyl persulfide, phenylacetyl disulfide 5g, s-2-phenylacetyl sulfanyl 2-phenylethanethioateShow MoreShow Less
PubChem CID 3378603
IUPAC Name S-(2-phenylacetyl)sulfanyl 2-phenylethanethioate
SMILES O=C(CC1=CC=CC=C1)SSC(=O)CC1=CC=CC=C1

Specifications

Melting Point 58°C to 60°C
Quantity 25 g
Beilstein 2057579
Solubility Information Insoluble in water.
Formula Weight 302.42
Percent Purity 98%
Chemical Name or Material Bis(phenylacetyl) disulfide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250296 Certificate TypeCertificate of Analysis Date10/04/2024 Catalog NumberL19792.14,L19792.06
Lot Number10248125 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog NumberL19792.14,L19792.06
Lot Number10237780 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL19792.14,L19792.06
Lot Number10245053 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog NumberL19792.14,L19792.06
Lot Number10242617 Certificate TypeCertificate of Analysis Date04/08/2022 Catalog NumberL19792.14,L19792.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(phenylacetyl) disulfide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.