missing translation for 'onlineSavingsMsg'
Learn More

Bis(pinacolato)diboron, 98+%, Thermo Scientific Chemicals

Product Code. 11302887
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Bis(pinacolato)diboron is a reagent used for the cis-vicinal diborylation of acetylenes and olefins with platinum catalysis. It acts as a substrate used in a new palladium-catalyzed cyclization of 1,6-enynes leading to homoallylic alkylboronates. It is used as a condensation agent in the preparation of poly(arylene)s. It plays an essential role in Suzuki reaction and used as matrix metallo-proteinase (MMP-2) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(pinacolato)diboron is a reagent used for the cis-vicinal diborylation of acetylenes and olefins with platinum catalysis. It acts as a substrate used in a new palladium-catalyzed cyclization of 1,6-enynes leading to homoallylic alkylboronates. It is used as a condensation agent in the preparation of poly(arylene)s. It plays an essential role in Suzuki reaction and used as matrix metallo-proteinase (MMP-2) inhibitor.

Solubility
Soluble in tetrahydrofuran, dichloromethane, toluene, hexane and heptane. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73183-34-3
Molecular Formula C12H24B2O4
Molecular Weight (g/mol) 253.94
MDL Number MFCD00799570
InChI Key IPWKHHSGDUIRAH-UHFFFAOYSA-N
Synonym bis pinacolato diboron, bis pinacolato diborane, 4,4,4',4',5,5,5',5'-octamethyl-2,2'-bi 1,3,2-dioxaborolane, pinacol diborane, diboron pinacol ester, b2pin2, 4,4,4',4',5,5,5',5'-octamethyl-2,2'-bi-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3,2-dioxaborolane, bispinacolate diboron, unii-i906w26p4u
PubChem CID 2733548
IUPAC Name 4,4,5,5-tetramethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 133°C to 139°C
Quantity 1 g
Beilstein 7703552
Solubility Information Soluble in tetrahydrofuran,dichloromethane,toluene,hexane and heptane. Insoluble in water.
Formula Weight 253.94
Percent Purity ≥98%
Chemical Name or Material Bis(pinacolato)diboron

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.