missing translation for 'onlineSavingsMsg'
Learn More

(+)-Bis[(R)-1-phenylethyl]amine, ChiPros™, 99%, ee 98+%, Thermo Scientific Chemicals

Product Code. 11397249
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(+)-Bis[(R)-1-phenylethyl]amine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-Bis[(R)-1-phenylethyl]amine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23294-41-9
Molecular Formula C16H19N
Molecular Weight (g/mol) 225.34
MDL Number MFCD00243088
InChI Key NXLACVVNHYIYJN-UHFFFAOYNA-N
Synonym +-bis r-1-phenylethyl amine, r-bis r-1-phenylethyl amine, bis r-alpha-methylbenzyl amine, +-bis r-1-phenylethy amine hydrochloride, 1r-1-phenyl-n-1r-1-phenylethyl ethanamine, bis 1r-1-phenylethyl amine, r,r-bis-1-phenylethyl amin, +-bis-r-1-phenylethyl amine hydrochloride
PubChem CID 5702599
IUPAC Name (1R)-1-phenyl-N-[(1R)-1-phenylethyl]ethanamine
SMILES CC(NC(C)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 98°C (0.3 mmHg)
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 225.34
Percent Purity 99%
Assay ee ≥98%
Chemical Name or Material (+)-Bis[(R)-1-phenylethyl]amine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'