Learn More
Bis(tri-n-butyltin) oxide, 97%, Thermo Scientific Chemicals
Description
Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.
Solubility
Soluble in water (0.071 g/L)
Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.

Specifications
Specifications
Melting Point | -45°C |
Density | 1.172 |
Boiling Point | 179°C to 180°C (2 mmHg) |
Flash Point | 168°C (334°F) |
Odor | Irritating |
Linear Formula | [(CH3CH2CH2CH2)3Sn]2O |
Refractive Index | 1.4865 |
Quantity | 100 g |
UN Number | UN2788 |
Beilstein | 745057 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Q26I066 | Certificate of Analysis | 02/05/2022 | A13242.0E,A13242.36,A13242.22 |
Q06I003 | Certificate of Analysis | 12/04/2022 | A13242.0E,A13242.36,A13242.22 |
Z06H008 | Certificate of Analysis | 12/04/2022 | A13242.0E,A13242.36,A13242.22 |
X20H035 | Certificate of Analysis | 12/04/2022 | A13242.0E,A13242.36,A13242.22 |
D5029C | Certificate of Analysis | 22/11/2021 | A13242.0E,A13242.36,A13242.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.