Learn More
Bis(trimethylsiloxy)methylsilane, 97%, Thermo Scientific Chemicals
£88.94 - £309.54
Chemical Identifiers
CAS | 1873-88-7 |
---|---|
Molecular Formula | C7H21O2Si3 |
Molecular Weight (g/mol) | 221.498 |
MDL Number | MFCD00048000 |
InChI Key | SWGZAKPJNWCPRY-UHFFFAOYSA-N |
Synonym | 1,1,1,3,5,5,5-heptamethyltrisiloxane, bis trimethylsiloxy methylsilane, unii-rvy9659r8c, trisiloxane, 1,1,1,3,5,5,5-heptamethyl, methyl-bis trimethylsilyloxy silicon, dsstox_cid_18795, methicone 20 cst, unii-6777u11mkt, methylpolysilicone, xiameter mhx-1107 fluid 20 cstShow More |
PubChem CID | 6327366 |
IUPAC Name | methyl-bis(trimethylsilyloxy)silicon |
SMILES | C[Si](O[Si](C)(C)C)O[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15457068
|
Thermo Scientific Alfa Aesar
H60586.AD |
50 mL |
£88.94
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15467068
|
Thermo Scientific Alfa Aesar
H60586.AK |
250 mL |
£309.54
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.
Solubility
Miscible with acetone, ethanol, diethyl ether. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
Chemical Identifiers
1873-88-7 | |
221.498 | |
SWGZAKPJNWCPRY-UHFFFAOYSA-N | |
6327366 | |
C[Si](O[Si](C)(C)C)O[Si](C)(C)C |
C7H21O2Si3 | |
MFCD00048000 | |
1,1,1,3,5,5,5-heptamethyltrisiloxane, bis trimethylsiloxy methylsilane, unii-rvy9659r8c, trisiloxane, 1,1,1,3,5,5,5-heptamethyl, methyl-bis trimethylsilyloxy silicon, dsstox_cid_18795, methicone 20 cst, unii-6777u11mkt, methylpolysilicone, xiameter mhx-1107 fluid 20 cstShow More | |
methyl-bis(trimethylsilyloxy)silicon |
Specifications
142°C | |
C7H21O2Si3 | |
MFCD00048000 | |
1748455 | |
1,1,1,3,5,5,5-heptamethyltrisiloxane, bis trimethylsiloxy methylsilane, unii-rvy9659r8c, trisiloxane, 1,1,1,3,5,5,5-heptamethyl, methyl-bis trimethylsilyloxy silicon, dsstox_cid_18795, methicone 20 cst, unii-6777u11mkt, methylpolysilicone, xiameter mhx-1107 fluid 20 cst | |
SWGZAKPJNWCPRY-UHFFFAOYSA-N | |
methyl-bis(trimethylsilyloxy)silicon | |
6327366 | |
97% | |
0.819 |
1873-88-7 | |
[(CH3)3SiO]2SiHCH3 | |
UN1993 | |
22°C (72°F) | |
Miscible with acetone,ethanol,diethyl ether. Insoluble in water. | |
C[Si](O[Si](C)(C)C)O[Si](C)(C)C | |
221.498 | |
222.51 | |
1.382 | |
Bis(trimethylsiloxy)methylsilane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 217-496-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.