missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)methane, 98%

Product Code. 11427587
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11427587

Brand: Thermo Scientific Alfa Aesar A15662.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2117-28-4 | C7H20Si2 | 160.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2117-28-4
Molecular Formula C7H20Si2
Molecular Weight (g/mol) 160.41
MDL Number MFCD00008275
InChI Key GYIODRUWWNNGPI-UHFFFAOYSA-N
Synonym bis trimethylsilyl methane, methylenebis trimethylsilane, silane, methylenebis trimethyl, trimethyl trimethylsilylmethyl silane, trimethyl trimethylsilyl methyl silane, 2,4-disilapentane, 2,2,4,4-tetramethyl, silane, 1,1'-methylenebis 1,1,1-trimethyl
PubChem CID 75029
IUPAC Name trimethyl(trimethylsilylmethyl)silane
SMILES C[Si](C)(C)C[Si](C)(C)C

Specifications

Boiling Point 132°C to 133°C
Melting Point -71°C
Quantity 1 g
UN Number UN1993
Beilstein 1734530
Flash Point 12°C (53°F)
Formula Weight 160.41
Percent Purity 98%
Refractive Index 1.417
Density 0.75
Chemical Name or Material Bis(trimethylsilyl)methane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.