Learn More
Bis(trimethylsilyl)sulfide, 98%, Thermo Scientific Chemicals
Description
Bis(trimethylsilyl)sulfide is used as a silylating agent in the synthesis of pseudohalides, trifluoroacetate and tetramethylsilyl halides. It is used in the transformation of oxides and chlorides into corresponding sulfides. It is also used in the preparation of dimethyltrisulfane as well as thiones from aldehydes and ketones. Further, it is used as a reducing agent to reduce aromatic nitro compounds to amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.851 g/mL |
Boiling Point | 162°C to 166°C |
Flash Point | 26°C (79°F) |
Odor | Pungent |
Refractive Index | 1.4598 |
Quantity | 1 g |
UN Number | UN1993 |
Beilstein | 1740046 |
Solubility Information | Miscible with terahydrofuran and toluene. |
Formula Weight | 178.44 |
Show More |
RUO – Research Use Only