missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)dicarbonylnickel, 98%, Thermo Scientific Chemicals

Product Code. 11323638
Click to view available options
Quantity:
2.5 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13007-90-4
Molecular Formula C38H30NiO2P2
Molecular Weight (g/mol) 639.30
MDL Number MFCD00009591
InChI Key SLFKPACCQUVAPG-UHFFFAOYSA-N
Synonym bis triphenylphosphine dicarbonylnickel
PubChem CID 22836333
IUPAC Name carbon monoxide;nickel;triphenylphosphanium
SMILES [Ni].[C-]#[O+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 210°C to 215°C (decomposition)
Quantity 2.5 g
UN Number UN3466
Merck Index 14,1304
Formula Weight 639.29
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Bis(triphenylphosphine)dicarbonylnickel

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'