missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismarck Brown Y, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Plasma stain for mucins, cartilage and goblet cellsBismarck Brown Y is used for staining mucine, cartilage and goblet cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Plasma stain for mucins, cartilage and goblet cellsBismarck Brown Y is used for staining mucine, cartilage and goblet cells.
Solubility
Soluble in water. Slightly soluble in ethanol and cellosolve. Insoluble in acetone, carbon tetrachloride and benzene.
Notes
Incompatible with strong oxidizing agents and reducing agents.
Plasma stain for mucins, cartilage and goblet cellsBismarck Brown Y is used for staining mucine, cartilage and goblet cells.
Solubility
Soluble in water. Slightly soluble in ethanol and cellosolve. Insoluble in acetone, carbon tetrachloride and benzene.
Notes
Incompatible with strong oxidizing agents and reducing agents.
Specifications
Merck Index | 14,1253 |
Solubility Information | Soluble in water; Slightly soluble in ethanol and cellosolve; Insoluble in acetone,carbon tetrachloride and benzene. |
Formula Weight | 419.32 |
Quantity | 25 g |
Chemical Name or Material | Bismarck Brown Y |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'