Learn More
Bismarck Brown Y, Thermo Scientific Chemicals
£19.30 - £40.00
Chemical Identifiers
CAS | 10114-58-6 |
---|---|
Molecular Formula | C18H20Cl2N8 |
Molecular Weight (g/mol) | 419.314 |
MDL Number | MFCD00012977 |
InChI Key | MCZVRBLCRZWFJH-UHFFFAOYSA-N |
Synonym | C.I. 21000 |
PubChem CID | 82360 |
ChEBI | CHEBI:53615 |
IUPAC Name | 4-[[3-[(2,4-diaminophenyl)diazenyl]phenyl]diazenyl]benzene-1,3-diamine;dihydrochloride |
SMILES | C1=CC(=CC(=C1)N=NC2=C(C=C(C=C2)N)N)N=NC3=C(C=C(C=C3)N)N.Cl.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11402668
|
Thermo Scientific Alfa Aesar
A16674.14 |
25 g |
£19.30
25g |
In Stock
Log in to see stock. |
|||||
11412668
|
Thermo Scientific Alfa Aesar
A16674.22 |
100 g |
£40.00
100g |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||||
Description
Plasma stain for mucins, cartilage and goblet cellsBismarck Brown Y is used for staining mucine, cartilage and goblet cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPlasma stain for mucins, cartilage and goblet cellsBismarck Brown Y is used for staining mucine, cartilage and goblet cells.
Solubility
Soluble in water. Slightly soluble in ethanol and cellosolve. Insoluble in acetone, carbon tetrachloride and benzene.
Notes
Incompatible with strong oxidizing agents and reducing agents.
Chemical Identifiers
10114-58-6 | |
419.314 | |
MCZVRBLCRZWFJH-UHFFFAOYSA-N | |
82360 | |
4-[[3-[(2,4-diaminophenyl)diazenyl]phenyl]diazenyl]benzene-1,3-diamine;dihydrochloride |
C18H20Cl2N8 | |
MFCD00012977 | |
C.I. 21000 | |
CHEBI:53615 | |
C1=CC(=CC(=C1)N=NC2=C(C=C(C=C2)N)N)N=NC3=C(C=C(C=C3)N)N.Cl.Cl |
Specifications
10114-58-6 | |
MFCD00012977 | |
C.I. 21000 | |
MCZVRBLCRZWFJH-UHFFFAOYSA-N | |
4-[[3-[(2,4-diaminophenyl)diazenyl]phenyl]diazenyl]benzene-1,3-diamine;dihydrochloride | |
82360 | |
419.32 |
C18H20Cl2N8 | |
14,1253 | |
Soluble in water; Slightly soluble in ethanol and cellosolve; Insoluble in acetone,carbon tetrachloride and benzene. | |
C1=CC(=CC(=C1)N=NC2=C(C=C(C=C2)N)N)N=NC3=C(C=C(C=C3)N)N.Cl.Cl | |
419.314 | |
CHEBI:53615 | |
Bismarck Brown Y |
Safety and Handling
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
H228-H302+H312+H332
missing translation for 'einecsNumber' : 233-314-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only