missing translation for 'onlineSavingsMsg'
Learn More

Bismuth antimonide, 99.99%

Product Code. 11488866
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11488866

Brand: Thermo Scientific Alfa Aesar A17014.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 12323-19-2 | Bi2Sb2 | 661.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12323-19-2
Molecular Formula Bi2Sb2
Molecular Weight (g/mol) 661.48
MDL Number MFCD00014196
InChI Key AEMQIQQWIVNHAU-UHFFFAOYSA-N

Specifications

Quantity 50 g
UN Number UN1549
Formula Weight 330.73
Percent Purity 99.99%
Chemical Name or Material Bismuth antimonide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.