missing translation for 'onlineSavingsMsg'
Learn More

Bismuth carbonate oxide, 98.5% min, Thermo Scientific Chemicals

Catalog Number 11327528
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
5892-10-4
CBi2O5
509.97
MFCD00010877
MGLUJXPJRXTKJM-UHFFFAOYSA-L
bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
16683095
CHEBI:31291
bis(oxobismuthanyl) carbonate
C(=O)(O[Bi]=O)O[Bi]=O
This item is not returnable. View return policy
5892-10-4
CBi2O5
509.97
MFCD00010877
MGLUJXPJRXTKJM-UHFFFAOYSA-L
bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
16683095
CHEBI:31291
bis(oxobismuthanyl) carbonate
C(=O)(O[Bi]=O)O[Bi]=O

Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes

Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazesBismuth carbonate oxide is used in ceramic glazes and pearly coatings on plastics. It is also used as an antacid and anti-diarrheal in medicinal chemistry. Further, it is used as a filler in radiopaque catheters. It is also employed in fireworks and to make Dragon's eggs. In addition to this, it is made into nanotube arrays that exhibit antibacterial properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 100 g
Merck Index 14,1282
Solubility Information Practically insoluble in water,alcohol; Soluble in nitric acid,hydrochloric acid,concentrated acetic acid;Soluble in mineral acid and glacial acetic acid; Insoluble in water.
Formula Weight 509.97
Percent Purity ≤98.5%
Density 6.86 g/mL
Chemical Name or Material Bismuth carbonate oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'