missing translation for 'onlineSavingsMsg'
Learn More

Bismuth carbonate oxide, Thermo Scientific Chemicals

£22.81 - £209.04

Chemical Identifiers

CAS 5892-10-4
Molecular Formula CBi2O5
Molecular Weight (g/mol) 509.97
MDL Number MFCD00010877
InChI Key MGLUJXPJRXTKJM-UHFFFAOYSA-L
Synonym bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
PubChem CID 16683095
ChEBI CHEBI:31291
IUPAC Name bis(oxobismuthanyl) carbonate
SMILES C(=O)(O[Bi]=O)O[Bi]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11434104
View Documents
Thermo Scientific Alfa Aesar
A12483.18
50 g
£22.81
50g
Please to purchase this item. Need a web account? Register with us today!
11444104
View Documents
Thermo Scientific Alfa Aesar
A12483.30
250 g
£65.22
250g
Please to purchase this item. Need a web account? Register with us today!
11424104
View Documents
Thermo Scientific Alfa Aesar
A12483.0B
1000 g
£209.04
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5892-10-4
509.97
MGLUJXPJRXTKJM-UHFFFAOYSA-L
16683095
bis(oxobismuthanyl) carbonate
CBi2O5
MFCD00010877
bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
CHEBI:31291
C(=O)(O[Bi]=O)O[Bi]=O

Specifications

5892-10-4
MFCD00010877
bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
C(=O)(O[Bi]=O)O[Bi]=O
509.97
CHEBI:31291
6.86
CBi2O5
14,1282
MGLUJXPJRXTKJM-UHFFFAOYSA-L
bis(oxobismuthanyl) carbonate
16683095
509.97
Bismuth carbonate oxide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 227-567-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only