missing translation for 'onlineSavingsMsg'
Learn More
Bismuth hydroxide, tech., Thermo Scientific Chemicals Product Code.: 11333988

Bismuth hydroxide, tech., Thermo Scientific Chemicals

Product Code. 11333988
1 kg
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11333988

Brand: Thermo Scientific Alfa Aesar 040397.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrolysis of ribonucleic acid, absorbent

It is used for the hydrolysis of ribonucleic acid and as absorbent for rutin and quercetin. It is a component used in milk of bismuth is used in gastrointestinal disorders as a protective agent. It is used for the removal of nitrate and other anions from water by yellow bismuth hydroxide. A bismuth oxide nanoplate is useful for carbon dioxide gas sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-43-0
Molecular Formula BiH3O3
Molecular Weight (g/mol) 260.00
MDL Number MFCD00046188
InChI Key TZSXPYWRDWEXHG-UHFFFAOYSA-K
IUPAC Name bismuth(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Bi+3]

Specifications

Physical Form Amorphous Powder
Quantity 1 kg
Solubility Information Insoluble in water; Soluble in acids.
Formula Weight 260
Grade Technical
Odor Odorless
Chemical Name or Material Bismuth hydroxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX30J036 Certificate TypeCertificate of Analysis Date15/05/2024 Catalog Number040397.30,040397.A1
Lot NumberX21G009 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog Number040397.30,040397.A1
Lot NumberQ14J013 Certificate TypeCertificate of Analysis Date26/04/2023 Catalog Number040397.A1
Lot NumberR11I036 Certificate TypeCertificate of Analysis Date02/08/2022 Catalog Number040397.30,040397.A1
Lot NumberM16B047 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040397.30,040397.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth hydroxide, tech., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.