missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth hydroxide, Thermo Scientific Chemicals
£29.70 - £107.00
Chemical Identifiers
CAS | 10361-43-0 |
---|---|
Molecular Formula | BiH3O3 |
Molecular Weight (g/mol) | 260.00 |
MDL Number | MFCD00046188 |
InChI Key | TZSXPYWRDWEXHG-UHFFFAOYSA-K |
IUPAC Name | bismuth(3+) trihydroxide |
SMILES | [OH-].[OH-].[OH-].[Bi+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10590574
|
Thermo Scientific Acros
393290500 |
50 g | Glass bottle |
£29.70
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10590764
|
Thermo Scientific Acros
393292500 |
250 g | Glass bottle |
£107.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10361-43-0 | |
260.00 | |
TZSXPYWRDWEXHG-UHFFFAOYSA-K | |
[OH-].[OH-].[OH-].[Bi+3] |
BiH3O3 | |
MFCD00046188 | |
bismuth(3+) trihydroxide |
Specifications
415.0°C | |
White to Cream | |
BiH3O3 | |
MFCD00046188 | |
Solubility in water: negligible. Other solubilities: soluble in acids | |
[OH-].[OH-].[OH-].[Bi+3] | |
260.00 | |
Positive | |
Bismuth hydroxide |
10361-43-0 | |
Powder | |
Bi(OH)3 | |
15, 1267 | |
TZSXPYWRDWEXHG-UHFFFAOYSA-K | |
bismuth(3+) trihydroxide | |
260 | |
1.0% max. |
Safety and Handling
missing translation for 'einecsNumber' : 233-790-2
RUO – Research Use Only