Learn More
Bismuth(III) chloride, 99.999%, (trace metal basis), Thermo Scientific Chemicals
£50.17 - £163.82
Chemical Identifiers
CAS | 7787-60-2 |
---|---|
Molecular Formula | BiCl3 |
Molecular Weight (g/mol) | 315.33 |
MDL Number | MFCD00003461 |
InChI Key | JHXKRIRFYBPWGE-UHFFFAOYSA-K |
Synonym | bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride |
PubChem CID | 24591 |
IUPAC Name | trichlorobismuthane |
SMILES | Cl[Bi](Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10185800
|
Thermo Scientific Acros
318350050 |
5 g | Glass bottle |
£50.17
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10561554
|
Thermo Scientific Acros
318350250 |
25 g | Glass bottle |
£163.82
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
7787-60-2 | |
315.33 | |
JHXKRIRFYBPWGE-UHFFFAOYSA-K | |
24591 | |
Cl[Bi](Cl)Cl |
BiCl3 | |
MFCD00003461 | |
bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride | |
trichlorobismuthane |
Specifications
447°C | |
7787-60-2 | |
Crystalline Powder and/or Crystals | |
BiCl3 | |
MFCD00003461 | |
15, 1264 | |
Solubility in water: decomposes. Other solubilities: soluble in acids,soluble in abs.alcohol,acetone and ethyl acetate | |
Cl[Bi](Cl)Cl | |
315.33 | |
315.33 | |
Trace Metal Basis | |
4.7500g/mL |
230°C to 232°C | |
White | |
99.999% | |
BiCl3 | |
07,24 | |
bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride | |
JHXKRIRFYBPWGE-UHFFFAOYSA-K | |
trichlorobismuthane | |
24591 | |
100.00% | |
4.75 | |
Bismuth(III) chloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 232-123-2
- Bismuth(III) chloride, anhydrous
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.