missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) chloride, ultra dry, 99.997+% (metals basis)

Product Code. 11353847
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
missing translation for 'unitSize'
25g
5g
This item is not returnable. View return policy

Product Code. 11353847

missing translation for 'mfr': Thermo Scientific Alfa Aesar 035680.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7787-60-2 | BiCl3 | 315.33 g/mol

Bismuth chloride is used as a catalyst in organic synthesis. In particular, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. The addition of other metal iodides increases its catalytic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-60-2
Molecular Formula BiCl3
Molecular Weight (g/mol) 315.33
MDL Number MFCD00003461
InChI Key JHXKRIRFYBPWGE-UHFFFAOYSA-K
Synonym bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride
PubChem CID 24591
IUPAC Name trichlorobismuthane
SMILES Cl[Bi](Cl)Cl

Specifications

Boiling Point ∼430°C (sublimation)
Melting Point 230°C to 232°C
Color White
Physical Form Powder
Quantity 5 g
UN Number UN3260
Merck Index 14,1262
Solubility Information It is soluble in methanol, diethyl ether, acetone.
Formula Weight 315.34
Percent Purity ≥99.997%
Packaging Ampule
Sensitivity Hygroscopic
Density 4.75 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth(III) chloride, Ultra dry
Show More Show Less
compliance-icons
Product Identifier
  • Bismuth(III) chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.