missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) fluoride, anhydrous, 99%, Thermo Scientific Chemicals

Manufacturing of bismuth pentafluoride

£34.87 - £337.68

Chemical Identifiers

CAS 7787-61-3
Molecular Formula BiF3
Molecular Weight (g/mol) 265.976
MDL Number MFCD00010893
InChI Key BRCWHGIUHLWZBK-UHFFFAOYSA-K
Synonym bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
PubChem CID 82233
IUPAC Name trifluorobismuthane
SMILES F[Bi](F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11372168
View Documents
Thermo Scientific Alfa Aesar
013075.14
25 g
£34.87
25g
Please to purchase this item. Need a web account? Register with us today!
11392168
View Documents
Thermo Scientific Alfa Aesar
013075.22
100 g
£91.25
100g
Please to purchase this item. Need a web account? Register with us today!
11382168
View Documents
Thermo Scientific Alfa Aesar
013075.36
500 g
£337.68
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in the manufacturing of bismuth pentafluoride. Bismuth(III) fluoride is an attractive reagent and catalyst in organic synthesis owing to its low cost, and ease of handling. It is also used in polarography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7787-61-3
265.976
BRCWHGIUHLWZBK-UHFFFAOYSA-K
82233
F[Bi](F)F
BiF3
MFCD00010893
bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
trifluorobismuthane

Specifications

727°C
Powder
MFCD00010893
14,1265
Insoluble in water; Soluble in HF.
F[Bi](F)F
265.976
265.98
Odorless
Hygroscopic
7787-61-3
BiF3
UN3260
bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
BRCWHGIUHLWZBK-UHFFFAOYSA-K
trifluorobismuthane
82233
99%
1.74
Bismuth(III) fluoride, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H319
Causes severe skin burns and eye damage.
Causes serious eye irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 232-124-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Bismuth(III) fluoride, anhydrous

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only