missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

£41.14 - £479.39

Chemical Identifiers

CAS 7787-64-6
Molecular Formula BiI3
Molecular Weight (g/mol) 589.69
MDL Number MFCD00010894
InChI Key KOECRLKKXSXCPB-UHFFFAOYSA-K
Synonym bismuth iodide, bismuth iii iodide, bismuth triiodide, bismuthine, triiodo, triiodobismuthine, triiodobismuth, bismuth iodide,, ksc179g5d
PubChem CID 111042
IUPAC Name bismuth(3+) triiodide
SMILES [I-].[I-].[I-].[Bi+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11393847
SDS View Documents
Thermo Scientific Alfa Aesar
035681.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11373847
SDS View Documents
Thermo Scientific Alfa Aesar
035681.06
5 g
£135.68
5g
Please to purchase this item. Need a web account? Register with us today!
11383847
SDS View Documents
Thermo Scientific Alfa Aesar
035681.14
25 g
£479.39
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth(III) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.