missing translation for 'onlineSavingsMsg'
Learn More
Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11380869

Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11380869
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11380869

Brand: Thermo Scientific Alfa Aesar 010657.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in electronic ceramics and fluorescent paints. It is employed as an oxidant for selective oxidation of sulfides to sulfoxides, as a nitrating reagent for synthesizing nitro aromatic compounds, and as a precursor for Dragendorff reagent for use in TLC stain and for testing the presence of alkaloids. On silica gel support, it has been reported to be an efficient catalyst for the Biginelli reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-44-1
Molecular Formula BiN3O9
Molecular Weight (g/mol) 394.99
MDL Number MFCD00003460
InChI Key PPNKDDZCLDMRHS-UHFFFAOYSA-N
Synonym bismuth nitrate, bismuth trinitrate, ccris 7110, nitric acid, bismuth 3+ salt, tris nitrooxy bismuthine, bismuth 3+ trinitrate, bi.3no3, bismuth iii nitrate, bismuth iii nitrate hydrate, nitric acid, bismuth 3+ salt 3:1
PubChem CID 107711
IUPAC Name bismuth;trinitrate
SMILES [Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 30°C (decomposition)
Color Colorless to White
Physical Form Crystalline
Quantity 25 g
UN Number UN1477
Merck Index 14,1271
Solubility Information Slightly soluble in water; soluble in acetone,acetic acid,and glycerol. Insoluble in ethyl acetate and ethanol.
Formula Weight 394.99 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic™
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Bismuth(III) nitrate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU23K052 Certificate TypeCertificate of Analysis Date17/09/2024 Catalog Number010657.22,010657.14
Lot NumberS04K029 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number010657.22,010657.14
Lot NumberW21J042 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number010657.22,010657.14
Lot NumberU28E049 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010657.22,010657.14
Lot Number24679 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010657.22,010657.14
compliance-icons compliance-icons
Product Identifier
  • Bismuth(III) nitrate hydrate, Puratronic
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.