Learn More
Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
£71.10 - £316.90
Chemical Identifiers
CAS | 10361-46-3 |
---|---|
Molecular Formula | BiNO4 |
Molecular Weight (g/mol) | 286.98 |
MDL Number | MFCD00064844 |
InChI Key | OGCYGKRWXWQOAK-UHFFFAOYSA-N |
Synonym | Bismuth oxynitrate |
IUPAC Name | (nitrooxy)bismuthanone |
SMILES | [O-][N+](=O)O[Bi]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11396138
|
Thermo Scientific Alfa Aesar
011122.09 |
10 g |
£71.10
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386138
|
Thermo Scientific Alfa Aesar
011122.18 |
50 g |
£305.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376138
|
Thermo Scientific Alfa Aesar
011122.30 |
250 g | |||||||
Description
Bismuth(III) nitrate oxide is used as an organic chemical synthesis intermediate. It is also used in the spectrophotometric detection of bismuth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10361-46-3 | |
286.98 | |
OGCYGKRWXWQOAK-UHFFFAOYSA-N | |
(nitrooxy)bismuthanone |
BiNO4 | |
MFCD00064844 | |
Bismuth oxynitrate | |
[O-][N+](=O)O[Bi]=O |
Specifications
10361-46-3 | |
BiNO4 | |
UN1477 | |
Insoluble in water. Soluble in acids. | |
[O-][N+](=O)O[Bi]=O | |
286.98 | |
99.998% | |
(metals basis) |
Powder | |
MFCD00064844 | |
Bismuth oxynitrate | |
OGCYGKRWXWQOAK-UHFFFAOYSA-N | |
(nitrooxy)bismuthanone | |
286.99 | |
Puratronic™ | |
Bismuth(III) nitrate oxide |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 233-792-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Bismuth nitrate oxide
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only