missing translation for 'onlineSavingsMsg'
Learn More
Bismuth(III) oxyfluoride, 97%, pure, Thermo Scientific™ Product Code.: 15249866

Bismuth(III) oxyfluoride, 97%, pure, Thermo Scientific™

Product Code. 15249866
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 15249866

Brand: Thermo Scientific Acros 453825000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13520-72-4
Molecular Formula BiFO
Molecular Weight (g/mol) 243.98

Specifications

Melting Point 500.0°C
Quantity 500 g
Assay Percent Range 96% min. (ex Bi)
Linear Formula FBi=O
Solubility Information Solubility in water: insoluble.
Formula Weight 243.98
Percent Purity 97%
Grade Pure
Chemical Name or Material Bismuth(III) oxyfluoride
compliance-icons compliance-icons
Product Identifier
  • Bismuth(III) oxyfluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth(III) oxyfluoride, 97%, pure, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.