missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) sulfate, 99%, pure, Thermo Scientific Chemicals
£28.44 - £79.29
Chemical Identifiers
CAS | 7787-68-0 |
---|---|
Molecular Formula | Bi2O12S3 |
Molecular Weight (g/mol) | 706.13 |
MDL Number | MFCD00036140 |
InChI Key | BEQZMQXCOWIHRY-UHFFFAOYSA-H |
Synonym | bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4 |
PubChem CID | 198144 |
IUPAC Name | dibismuth;trisulfate |
SMILES | [Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15209876
|
Thermo Scientific Acros
453850250 |
25 g |
£28.44
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15219876
|
Thermo Scientific Acros
453851000 |
100 g |
£79.29
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7787-68-0 | |
706.13 | |
BEQZMQXCOWIHRY-UHFFFAOYSA-H | |
198144 | |
[Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Bi2O12S3 | |
MFCD00036140 | |
bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4 | |
dibismuth;trisulfate |
Specifications
405.0°C | |
98.5% min | |
Bi2(SO4)3 | |
15,1286 | |
BEQZMQXCOWIHRY-UHFFFAOYSA-H | |
dibismuth;trisulfate | |
198144 | |
99% | |
Bismuth(III) sulfate |
7787-68-0 | |
Bi2O12S3 | |
MFCD00036140 | |
bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4 | |
[Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
706.13 | |
706.13 | |
Pure |
Safety and Handling
missing translation for 'einecsNumber' : 232-129-5
Product Identifier
- Bismuth(III) sulfate
RUO – Research Use Only