Learn More
Bismuth(III) sulfate, 99%, Thermo Scientific Chemicals
£39.40 - £123.62
Chemical Identifiers
CAS | 7787-68-0 |
---|---|
Molecular Formula | Bi2O12S3 |
Molecular Weight (g/mol) | 706.13 |
MDL Number | MFCD00036140 |
InChI Key | BEQZMQXCOWIHRY-UHFFFAOYSA-H |
Synonym | bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4 |
PubChem CID | 198144 |
IUPAC Name | dibismuth;trisulfate |
SMILES | [Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15433237
|
Thermo Scientific Alfa Aesar
B22007.14 |
25 g |
£39.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15443237
|
Thermo Scientific Alfa Aesar
B22007.22 |
100 g |
£123.62
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The esterification of thujopsene with bismuth(III) sulfate in carboxylic acids yeilds esters in 30%. Bismuth(III) sulfateis used as analytics reagents and medicine, it is also used in pharmaceutical industry. It is used to determine other metal sulphate salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe esterification of thujopsene with bismuth(III) sulfate in carboxylic acids yeilds esters in 30%. Bismuth(III) sulfateis used as analytics reagents and medicine, it is also used in pharmaceutical industry. It is used to determine other metal sulphate salts.
Solubility
Soluble in acid, insoluble in water and ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
7787-68-0 | |
706.13 | |
BEQZMQXCOWIHRY-UHFFFAOYSA-H | |
198144 | |
[Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Bi2O12S3 | |
MFCD00036140 | |
bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4 | |
dibismuth;trisulfate |
Specifications
405°C (decomposition) | |
Bi2O12S3 | |
MFCD00036140 | |
bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4 | |
BEQZMQXCOWIHRY-UHFFFAOYSA-H | |
dibismuth;trisulfate | |
198144 | |
99% | |
Bismuth(III) sulfate |
7787-68-0 | |
Bi2(SO4)3 | |
14,1286 | |
Soluble in acid,insoluble in water and ethanol. | |
[Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
706.13 | |
706.13 | |
5.08 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 232-129-5
missing translation for 'rtecsNumber' : WS6600000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Bismuth(III) sulfate
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only