missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) telluride, Vacuum Deposition Grade, 99.999% (metals basis), Thermo Scientific Chemicals

£82.21 - £295.47

Chemical Identifiers

CAS 1304-82-1
MDL Number MFCD00014201
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11392468
View Documents
Thermo Scientific Alfa Aesar
014519.09
10 g
£82.21
10g
Please to purchase this item. Need a web account? Register with us today!
11302478
View Documents
Thermo Scientific Alfa Aesar
014519.18
50 g
£295.47
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bismuth(III) telluride and its alloys (antimony or selenium) posses excellent semiconductor activity, which is used in thermoelectric industry for cooling and energy generation. It increases the speed of microchips and acts as topological insulators in spintronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1304-82-1
MFCD00014201

Specifications

573°C
Gray
MFCD00014201
Insoluble in water.
99.999%
Moisture sensitive
Bismuth(III) telluride
1304-82-1
Pieces
UN3284
800.76
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TELLURIUM COMPOUND, N.O.S.

missing translation for 'einecsNumber' : 215-135-2

missing translation for 'rtecsNumber' : EB3110000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Bismuth(III) telluride, Vacuum Deposition Grade

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only