missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) trifluoromethanesulfonate, 98%

Product Code. 10124823 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10124823

Brand: Thermo Scientific Acros 387590050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88189-03-1 | C3BiF9O9S3 | 656.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88189-03-1
Molecular Formula C3BiF9O9S3
Molecular Weight (g/mol) 656.19
MDL Number MFCD02093669
InChI Key NYENCOMLZDQKNH-UHFFFAOYSA-K
Synonym bismuth iii trifluoromethanesulfonate, bismuth triflate, bismuth trifluoromethanesulfonate, bismuth 3+ triflate, bismuth triflate mi, bismuth 3+ trifluoromethanesulfonate, bismuth tris trifluoromethanesulfonate, bismuth iii triflate, bismuth 3+ ion tritriflate
PubChem CID 9917655
IUPAC Name bismuth;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Bi+3]

Specifications

Melting Point >300.0°C
Color Colorless
Physical Form Liquid
Quantity 5 g
Assay Percent Range 31.0 to 33% Bi (Complexometry)
Linear Formula Bi(OS(O2)CF3)3
Merck Index 15, 1292
Formula Weight 656.19
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Bismuth(III) trifluoromethanesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Bismuth(III) trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.