missing translation for 'onlineSavingsMsg'
Learn More
Bismuth(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals Product Code.: 10166763

Bismuth(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Product Code. 10166763
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10166763

Brand: Thermo Scientific Acros 387590250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88189-03-1
Molecular Formula C3BiF9O9S3
Molecular Weight (g/mol) 656.19
MDL Number MFCD02093669
InChI Key NYENCOMLZDQKNH-UHFFFAOYSA-K
Synonym bismuth iii trifluoromethanesulfonate, bismuth triflate, bismuth trifluoromethanesulfonate, bismuth 3+ triflate, bismuth triflate mi, bismuth 3+ trifluoromethanesulfonate, bismuth tris trifluoromethanesulfonate, bismuth iii triflate, bismuth 3+ ion tritriflate
PubChem CID 9917655
IUPAC Name bismuth;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Bi+3]

Specifications

Melting Point >300.0°C
Color Colorless
Physical Form Liquid
Quantity 25 g
Assay Percent Range 31.0 to 33% Bi (Complexometry)
Linear Formula Bi(OS(O2)CF3)3
Merck Index 15, 1292
Formula Weight 656.19
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Bismuth(III) trifluoromethanesulfonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468769 Certificate TypeCertificate of Analysis Date17/01/2025 Catalog Number387590050,387590250
Lot NumberA0465566 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number387590050,387590250
Lot NumberA0458195 Certificate TypeCertificate of Analysis Date06/12/2023 Catalog Number387590050,387590250
Lot NumberA0456950 Certificate TypeCertificate of Analysis Date23/10/2023 Catalog Number387590050,387590250
Lot NumberA0455021 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number387590050,387590250
compliance-icons
Product Identifier
  • Bismuth(III) trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.