missing translation for 'onlineSavingsMsg'
Learn More
Bismuth ingot/button, ^=36mm (1.4in) dia x 8mm (0.3in) thick, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 15404945

Bismuth ingot/button, ^=36mm (1.4in) dia x 8mm (0.3in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15404945
1 Pc.
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15404945

Brand: Thermo Scientific Alfa Aesar 045102.KF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
CAS 7440-69-9
Molecular Formula Bi
MDL Number MFCD00134033
Quantity 1 Pc.
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
SMILES [Bi]
IUPAC Name bismuth
Molecular Weight (g/mol) 208.98
PubChem CID 5359367
ChEBI CHEBI:33301
Percent Purity 99.9%
Odor Odorless
Weight ≈60g/piece
Form Pieces
Assay (metals basis)
Chemical Name or Material Bismuth ingot/button, ≈36mm (1.4 in.) dia. x 8mm (0.3 in.) thick
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth ingot/button, ^=36mm (1.4in) dia x 8mm (0.3in) thick, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.