missing translation for 'onlineSavingsMsg'
Learn More
Bismuth Lead Tin Cadmium ingot (Wood's metal), Thermo Scientific Chemicals Product Code.: 11357528

Bismuth Lead Tin Cadmium ingot (Wood's metal), Thermo Scientific Chemicals

Product Code. 11357528
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11357528

Brand: Thermo Scientific Alfa Aesar 033218.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Wood's metal is useful as a low-melting solder, low-temperature casting metal, high temperature coupling fluid in heat baths, and as a fire-melted valve element in fire sprinkler systems in buildings. It is also used as a filler when bending thin-walled metal tube.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8049-22-7
Molecular Formula Cd12Sn2
Molecular Weight (g/mol) 1586.388
MDL Number MFCD00198175
InChI Key OIBLPQFTAJDRMC-UHFFFAOYSA-N
Synonym lambda∼2∼-stannane-cadmium 1/6
PubChem CID 71434152
IUPAC Name cadmium;tin
SMILES [Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Sn].[Sn]

Specifications

Color Gray
Melting Point 71°C
Avg. Mol. Wt. or Mol. Wt. Range Bi:Pb:Sn:Cd; 50:25:12.5:12.5 wt%
UN Number UN3077
Quantity 100 g
Solubility Information Insoluble in water.
Odor Odorless
Form Ingot
Assay (Wood's metal)
Chemical Name or Material Bismuth Lead Tin Cadmium ingot (Wood's metal)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR31K032 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number033218.36,033218.22,033218.A3
Lot NumberP16I004 Certificate TypeCertificate of Analysis Date22/06/2022 Catalog Number033218.36,033218.22,033218.A3
Lot NumberI19Q76 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number033218.36,033218.22,033218.A3
Lot NumberK20Z082 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number033218.36,033218.22,033218.A3
Lot NumberU07D010 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number033218.36,033218.22,033218.A3
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bismuth Lead Tin Cadmium ingot (Wood's metal)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H372-Causes damage to organs.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P201-Obtain special instructions before use.
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Lead, Cadmium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth Lead Tin Cadmium ingot (Wood's metal), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.